Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmed1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013432.1 | NP_001013450.1 | 3 - 4 | 467 | 65 | |
XM_006242644.1 | XP_006242706.1 | 4 - 5 | 1946 | 65 | ||
XM_008765940.1 | XP_008764162.1 | 4 - 5 | 1881 | 65 | ||
GenBank mRNA | BC091167.1 | - | 3 - 4 | 467 | 7324543 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |