Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012864.2 | NP_036996.1 | 5 - 6 | 805 | 81 | |
GenBank mRNA | BC064657.1 | - | 5 - 6 | 805 | 5623517 | 81 |
L24374.1 | - | 5 - 6 | 819 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease 3' Most Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |