Search
Search

Proser2| Gene Symbol: Proser2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008771897.1 | XP_008770119.1 | 2 - 3 | 415 | 66 | |
| XM_008771898.1 | XP_008770120.1 | 2 - 3 | 643 | 66 | ||
| XM_008771899.1 | XP_008770121.1 | 2 - 3 | 412 | 66 | ||
Set Membership: |
|