Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abhd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108866.1 | NP_001102336.1 | 3 - 4 | 556 | 106 | |
XM_006251995.2 | XP_006252057.1 | 3 - 4 | 761 | 106 | ||
XM_006251996.2 | XP_006252058.1 | 3 - 4 | 552 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |