Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Khnyn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751468.3 | XP_003751516.1 | 4 - 5 | 1718 | 62 | |
XM_006252032.2 | XP_006252094.1 | 4 - 5 | 1913 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |