Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elovl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191844.1 | NP_001178773.1 | 641 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Probe spans exons |