Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mier3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168000.1 | NP_001161472.1 | 3 - 4 | 403 | 122 | |
XM_006231924.2 | XP_006231986.1 | 5 - 6 | 498 | 122 | ||
XM_006231925.2 | XP_006231987.2 | 5 - 6 | 905 | 122 | ||
GenBank mRNA | JQ013732.1 | - | 3 - 4 | 1349 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |