Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gzmk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017119.2 | NP_058815.1 | 1 - 2 | 80 | 114 | |
GenBank mRNA | L19694.1 | - | 1 - 2 | 80 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Detect Genomic DNA Rattus norvegicus Probe spans exons |