Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008372.1 | NP_001008373.1 | 4 - 5 | 797 | 157 | |
XM_006231776.2 | XP_006231838.1 | 4 - 5 | 787 | 157 | ||
XM_006231778.2 | XP_006231840.1 | 4 - 5 | 836 | 157 | ||
XM_008760639.1 | XP_008758861.1 | 5 - 6 | 900 | 157 | ||
GenBank mRNA | BC085771.1 | - | 4 - 5 | 797 | 7191068 | 157 |
FQ211725.1 | - | 4 - 5 | 828 | 157 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |