Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dstyk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199463.2 | NP_955750.1 | 8 - 9 | 2163 | 98 | |
XM_006249774.2 | XP_006249836.1 | 8 - 9 | 2013 | 98 | ||
GenBank mRNA | AY208851.1 | - | 8 - 9 | 2094 | 98 | |
AY429675.1 | - | 8 - 9 | 2163 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons |