Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006249723.2 | XP_006249785.1 | 9 - 10 | 1163 | 144 | |
GenBank mRNA | AB026904.1 | - | 7 - 8 | 973 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |