Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769512.1 | XP_008767734.1 | 6 - 7 | 680 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons |