Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chi3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053560.1 | NP_446012.1 | 8 - 9 | 954 | 73 | |
XM_006249892.2 | XP_006249954.2 | 8 - 9 | 1008 | 73 | ||
XM_008769574.1 | XP_008767796.1 | 8 - 9 | 975 | 73 | ||
GenBank mRNA | AF062038.1 | - | 6 - 7 | 835 | 73 | |
BC091365.1 | - | 8 - 9 | 954 | 7319171 | 73 | |
EF467168.1 | - | 6 - 7 | 835 | 73 | ||
FQ232458.1 | - | 5 - 6 | 620 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |