Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcrlb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191931.1 | NP_001178860.1 | 4 - 5 | 570 | 76 | |
XM_008769747.1 | XP_008767969.1 | 2 - 3 | 754 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons |