Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107174.1 | NP_001100644.1 | 6 - 7 | 997 | 103 | |
XM_006249775.2 | XP_006249837.1 | 6 - 7 | 785 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |