Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133311.1 | NP_579845.1 | 4 - 5 | 620 | 90 | |
XM_008769421.1 | XP_008767643.1 | 4 - 5 | 772 | 90 | ||
GenBank mRNA | AF004774.2 | - | 4 - 5 | 569 | 90 | |
AF269251.1 | - | 4 - 5 | 620 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |