Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008364.1 | NP_001008365.1 | 5 - 6 | 778 | 88 | |
XM_006254726.2 | XP_006254788.1 | 6 - 7 | 825 | 88 | ||
XM_006254728.1 | XP_006254790.1 | 5 - 6 | 533 | 88 | ||
XM_006254729.2 | XP_006254791.1 | 6 - 7 | 403 | 88 | ||
XM_008772152.1 | XP_008770374.1 | 6 - 7 | 825 | 88 | ||
GenBank mRNA | BC085883.1 | - | 5 - 6 | 778 | 7192093 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |