Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106135.2 | NP_001099605.2 | 6 - 7 | 676 | 74 | |
XM_006254716.2 | XP_006254778.1 | 6 - 7 | 702 | 74 | ||
GenBank mRNA | BC166853.1 | - | 6 - 7 | 651 | 9092757 | 74 |
CB557295.1 | - | 4 - 5 | 280 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |