Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019314.1 | NP_062187.1 | 6 - 7 | 1381 | 85 | |
GenBank mRNA | U69882.1 | - | 6 - 7 | 1381 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Rattus norvegicus Probe spans exons |