Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slmo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109570.1 | NP_001103040.1 | 5 - 6 | 510 | 72 | |
XM_006254885.2 | XP_006254947.1 | 5 - 6 | 862 | 72 | ||
XM_006254886.2 | XP_006254948.1 | 5 - 6 | 863 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Rattus norvegicus Probe spans exons |