Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm7sf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011970.1 | NP_001011970.1 | 3 - 4 | 473 | 71 | |
GenBank mRNA | BC090030.1 | - | 3 - 4 | 473 | 7366240 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |