Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013091.1 | NP_037223.1 | 9 - 10 | 1315 | 70 | |
XM_006237334.2 | XP_006237396.1 | 8 - 9 | 1550 | 70 | ||
GenBank mRNA | AF329976.1 | - | 9 - 10 | 1314 | 70 | |
AF329977.1 | - | 9 - 10 | 1314 | 70 | ||
AF329978.1 | - | 9 - 10 | 1314 | 70 | ||
AF329979.1 | - | 9 - 10 | 1314 | 70 | ||
AF329980.1 | - | 9 - 10 | 1314 | 70 | ||
AF329981.1 | - | 9 - 10 | 1314 | 70 | ||
BC086413.1 | - | 9 - 10 | 1335 | 7110094 | 70 | |
M63122.1 | - | 9 - 10 | 1315 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons |