Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lag3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212513.2 | NP_997678.2 | 2 - 3 | 330 | 56 | |
XM_006237348.2 | XP_006237410.1 | 2 - 3 | 555 | 56 | ||
XM_008763282.1 | XP_008761504.1 | 3 - 4 | 436 | 56 | ||
GenBank mRNA | AY230414.1 | - | 2 - 3 | 220 | 56 | |
BC091201.1 | - | 2 - 3 | 330 | 7380279 | 56 | |
DQ438937.1 | - | 2 - 3 | 208 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |