Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191752.1 | NP_001178681.1 | 5 - 6 | 582 | 92 | |
XM_006237236.2 | XP_006237298.1 | 6 - 7 | 828 | 92 | ||
XM_006237237.2 | XP_006237299.1 | 6 - 7 | 816 | 92 | ||
XM_006237238.1 | XP_006237300.1 | 7 - 8 | 871 | 92 | ||
XM_006237239.1 | XP_006237301.1 | 7 - 8 | 853 | 92 | ||
XM_006237240.2 | XP_006237302.1 | 6 - 7 | 1739 | 92 | ||
XM_006237241.1 | XP_006237303.1 | 6 - 7 | 674 | 92 | ||
XM_006237242.2 | XP_006237304.1 | 5 - 6 | 1759 | 92 | ||
XM_006237243.2 | XP_006237305.1 | 5 - 6 | 1685 | 92 | ||
XM_006237244.2 | XP_006237306.1 | 5 - 6 | 582 | 92 | ||
XM_008763244.1 | XP_008761466.1 | 6 - 7 | 828 | 92 | ||
XM_008763245.1 | XP_008761467.1 | 6 - 7 | 1813 | 92 | ||
XM_008763246.1 | XP_008761468.1 | 6 - 7 | 669 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |