Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134705.1 | NP_001128177.1 | 1 - 2 | 220 | 66 | |
GenBank mRNA | BC166894.1 | - | 1 - 2 | 184 | 6920183 | 66 |
BI289576.1 | - | 2 - 3 | 66 | |||
BQ779631.1 | - | 2 - 3 | 66 | |||
FQ217721.1 | - | 1 - 2 | 169 | 66 | ||
FQ217795.1 | - | 1 - 2 | 169 | 66 | ||
FQ221226.1 | - | 1 - 2 | 230 | 66 | ||
FQ223619.1 | - | 1 - 2 | 171 | 66 | ||
FQ224715.1 | - | 1 - 2 | 199 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |