Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgcs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017268.1 | NP_058964.1 | 3 - 4 | 534 | 68 | |
XM_006231959.1 | XP_006232021.1 | 2 - 3 | 703 | 68 | ||
GenBank mRNA | AY724522.1 | - | 2 - 3 | 520 | 68 | |
FQ211444.1 | - | 2 - 3 | 506 | 68 | ||
X52625.1 | - | 3 - 4 | 534 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |