Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exosc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106432.1 | NP_001099902.1 | 4 - 5 | 696 | 67 | |
XM_006232332.2 | XP_006232394.1 | 5 - 6 | 277 | 67 | ||
GenBank mRNA | FQ230358.1 | - | 4 - 5 | 224 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |