Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033665.1 | NP_001028837.1 | 5 - 6 | 1015 | 122 | |
NM_012916.2 | NP_037048.2 | 5 - 6 | 1015 | 122 | ||
XM_006232595.2 | XP_006232657.1 | 5 - 6 | 2096 | 122 | ||
XM_006232596.2 | XP_006232658.1 | 5 - 6 | 999 | 122 | ||
XM_006232597.2 | XP_006232659.1 | 5 - 6 | 817 | 122 | ||
GenBank mRNA | BC094307.1 | - | 1 - 2 | 1795 | 122 | |
U37142.1 | - | 5 - 6 | 819 | 122 | ||
X79881.1 | - | 5 - 6 | 869 | 122 | ||
X86406.1 | - | 5 - 6 | 869 | 122 | ||
Z28366.1 | - | 5 - 6 | 1010 | 122 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |