Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247437.1 | XP_006247499.1 | 13 - 14 | 2532 | 50 | |
XM_006247438.1 | XP_006247500.1 | 13 - 14 | 2532 | 50 | ||
XM_008768198.1 | XP_008766420.1 | 11 - 12 | 2836 | 50 | ||
XM_008768199.1 | XP_008766421.1 | 12 - 13 | 2743 | 50 | ||
XM_008768200.1 | XP_008766422.1 | 11 - 12 | 3987 | 50 | ||
XM_220918.8 | XP_220918.6 | 15 - 16 | 2836 | 50 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |