Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcgf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105836.2 | NP_001099306.1 | 3 - 4 | 396 | 64 | |
XM_008767971.1 | XP_008766193.1 | 4 - 5 | 355 | 64 | ||
XM_008767972.1 | XP_008766194.1 | 4 - 5 | 631 | 64 | ||
XM_008767973.1 | XP_008766195.1 | 4 - 5 | 701 | 64 | ||
GenBank mRNA | BC167059.1 | - | 3 - 4 | 396 | 7445511 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |