Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcgf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105836.2 | NP_001099306.1 | 5 - 6 | 554 | 65 | |
XM_008767971.1 | XP_008766193.1 | 6 - 7 | 513 | 65 | ||
XM_008767972.1 | XP_008766194.1 | 6 - 7 | 789 | 65 | ||
XM_008767973.1 | XP_008766195.1 | 6 - 7 | 859 | 65 | ||
GenBank mRNA | BC167059.1 | - | 5 - 6 | 554 | 7445511 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |