Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005875.1 | NP_001005875.1 | 8 - 9 | 963 | 76 | |
GenBank mRNA | BC083758.1 | - | 8 - 9 | 963 | 7192849 | 76 |
FQ218292.1 | - | 2 - 3 | 190 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |