Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247855.2 | XP_006247917.1 | 3 - 4 | 428 | 103 | |
XM_221142.8 | XP_221142.5 | 3 - 4 | 428 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |