Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005874.2 | NP_001005874.2 | 1 | 2138 | 132 | |
GenBank mRNA | BC083841.1 | - | 1 | 2135 | 7114321 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Both primers and probe map within a single exon |