Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ell2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760603.1 | XP_008758825.1 | 11 - 12 | 2539 | 160 | |
XM_008760604.1 | XP_008758826.1 | 11 - 12 | 1825 | 160 | ||
GenBank mRNA | BC111075.1 | - | 2 - 3 | 151 | 160 | |
FQ230057.1 | - | 4 - 5 | 533 | 160 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |