Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053719.1 | NP_446171.1 | 6 | 965 | 131 | |
GenBank mRNA | AJ009698.1 | - | 6 | 965 | 131 | |
BC061846.1 | - | 6 | 967 | 5621074 | 131 | |
FQ213962.1 | - | 6 | 978 | 131 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Amplicon spans exons and probe does not span exons |