Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006230649.2 | XP_006230711.1 | 1 - 2 | 344 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |