Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749030.3 | XP_003749078.1 | 5 - 6 | 3213 | 80 | |
XM_006230921.2 | XP_006230983.1 | 6 - 7 | 2722 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons |