Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnmal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006228440.2 | XP_006228502.1 | 2 - 3 | 745 | 56 | |
XM_008758951.1 | XP_008757173.1 | 3 - 4 | 461 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |