Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snrnp35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014127.1 | NP_001014149.1 | 1 - 2 | 93 | 129 | |
GenBank mRNA | BC086435.1 | - | 1 - 2 | 93 | 7302221 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |