Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbscr22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135743.1 | NP_001129215.1 | 6 - 7 | 476 | 93 | |
GenBank mRNA | BC160891.1 | - | 6 - 7 | 476 | 7442902 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |