Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009715.1 | NP_001009715.1 | 10 - 11 | 2177 | 110 | |
GenBank mRNA | AY230746.1 | - | 10 - 11 | 2177 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus Inventoried Probe spans exons |