Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nobox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192013.1 | NP_001178942.1 | 5 - 6 | 644 | 134 | |
XM_008762912.1 | XP_008761134.1 | 5 - 6 | 839 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |