Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008762751.1 | XP_008760973.1 | 15 - 16 | 2897 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |