Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glcci1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177442.1 | NP_001170913.1 | 3 - 4 | 241 | 95 | |
GenBank mRNA | BC083591.1 | - | 3 - 4 | 267 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |