Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749716.3 | XP_003749764.1 | 9 - 10 | 1918 | 120 | |
XM_006236346.2 | XP_006236408.1 | 9 - 10 | 1918 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |