Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zyx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053761.1 | NP_446213.1 | 5 - 6 | 1188 | 73 | |
XM_006236387.2 | XP_006236449.1 | 5 - 6 | 1343 | 73 | ||
XM_006236388.2 | XP_006236450.1 | 5 - 6 | 1306 | 73 | ||
XM_006236389.2 | XP_006236451.1 | 4 - 5 | 1250 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |