Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Napg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107384.1 | NP_001100854.1 | 7 - 8 | 617 | 162 | |
XM_006254829.2 | XP_006254891.1 | 8 - 9 | 609 | 162 | ||
GenBank mRNA | AY724541.1 | - | 8 - 9 | 514 | 162 | |
FQ211390.1 | - | 5 - 6 | 491 | 162 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |