Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flrt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109160.1 | NP_001102630.1 | 1 | 5265 | 113 | |
XM_008760167.1 | XP_008758389.1 | 2 | 8598 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon |