Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glrx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032614.2 | NP_116003.2 | 3 - 4 | 285 | 95 | |
XM_006230497.2 | XP_006230559.1 | 3 - 4 | 698 | 95 | ||
GenBank mRNA | BC086381.1 | - | 3 - 4 | 285 | 7301390 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |